siRNA Details
| Gene Symbol: NEK4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001193533.1 | NP_001180462.1 | 13 | 2137 | ||
| NM_003157.4 | NP_003148.2 | 14 | 2404 | |||
| XM_006713310.2 | XP_006713373.1 | 13 | 2263 | |||
| XM_011534039.2 | XP_011532341.1 | 14 | 2401 | |||
| XM_017007085.1 | XP_016862574.1 | 14 | 2401 | |||
| XM_017007086.1 | XP_016862575.1 | 13 | 2263 | |||
| GenBank mRNA | AK294165.1 | - | 13 | 2044 | ||
| AK312420.1 | - | 14 | 2392 | |||
| BC063044.1 | - | 13 | 2256 | 5169184 | ||
| EF560744.1 | - | 14 | 2400 | |||
| KJ592714.1 | - | 13 | 2063 | |||
| L20321.1 | - | 14 | 2379 | |||

NEK4
Genomic Map