siRNA Details
| Gene Symbol: NEK4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001193533.1 | NP_001180462.1 | 13, 14 | 2233 | ||
| NM_003157.4 | NP_003148.2 | 14, 15 | 2500 | |||
| XM_006713310.2 | XP_006713373.1 | 13, 14 | 2359 | |||
| XM_011534039.2 | XP_011532341.1 | 14, 15 | 2497 | |||
| XM_017007085.1 | XP_016862574.1 | 14, 15 | 2497 | |||
| XM_017007086.1 | XP_016862575.1 | 13, 14 | 2359 | |||
| GenBank mRNA | AK294165.1 | - | 13, 14 | 2140 | ||
| AK312420.1 | - | 14, 15 | 2488 | |||
| BC063044.1 | - | 13, 14 | 2352 | 5169184 | ||
| EF560744.1 | - | 14, 15 | 2496 | |||
| L20321.1 | - | 14, 15 | 2475 | |||

NEK4
Genomic Map