siRNA Details
| Gene Symbol: EPHA2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | BC008655.1 | - | 5 | 1414 | ||
| BC037166.2 | - | 17 | 3700 | 5090654 | ||
| M59371.1 | - | 17 | 3695 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

EPHA2| Gene Symbol: EPHA2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | BC008655.1 | - | 5 | 1414 | ||
| BC037166.2 | - | 17 | 3700 | 5090654 | ||
| M59371.1 | - | 17 | 3695 | |||