siRNA Details
| Gene Symbol: PRKD3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_005813.4 | NP_005804.1 | 3, 4 | 1107 | ||
| XM_005264237.3 | XP_005264294.1 | 4, 5 | 1380 | |||
| XM_005264238.3 | XP_005264295.1 | 3, 4 | 356 | |||
| XM_017003741.1 | XP_016859230.1 | 5, 6 | 1385 | |||
| XM_017003742.1 | XP_016859231.1 | 4, 5 | 424 | |||
| GenBank mRNA | AB015982.2 | - | 3, 4 | 1107 | ||
| BC012820.2 | - | 3, 4 | 1066 | |||
| BC030706.2 | - | 3, 4 | 1052 | 5266769 | ||

PRKD3
Genomic Map