siRNA Details
Gene Symbol: ZNF107 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001013746.2 | NP_001013768.1 | 5 | 4434 | ||
NM_001282359.1 | NP_001269288.1 | 4 | 4274 | |||
NM_001282360.1 | NP_001269289.1 | 3 | 4178 | |||
NM_016220.4 | NP_057304.1 | 7 | 4703 | |||
XM_017012283.1 | XP_016867772.1 | 7 | 4859 | |||
XM_017012284.1 | XP_016867773.1 | 6 | 4732 | |||
XM_017012285.1 | XP_016867774.1 | 8 | 5001 | |||
XM_017012286.1 | XP_016867775.1 | 4 | 4296 | |||
GenBank mRNA | AB027251.1 | - | 4 | 5244 | ||
AK303931.1 | - | 6 | 1397 | |||
BC047243.1 | - | 5 | 4320 | 6066829 |
