siRNA Details
| Gene Symbol: SRPK3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001170760.1 | NP_001164231.1 | 5 | 513 | ||
| NM_001170761.1 | NP_001164232.1 | 5 | 513 | |||
| NM_014370.3 | NP_055185.2 | 5 | 513 | |||
| GenBank mRNA | AF027406.1 | - | 5 | 451 | ||
| AK301749.1 | - | 5 | 473 | |||
| AK314828.1 | - | 5 | 473 | |||
| BC092416.1 | - | 5 | 459 | 30529915 | ||
| BC117124.1 | - | 5 | 507 | 40125675 | ||
| DQ099381.1 | - | 5 | 427 | |||
| U82808.1 | - | 5 | 331 | |||

SRPK3
Genomic Map