siRNA Details
| Gene Symbol: NUAK2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_030952.2 | NP_112214.2 | 7 | 2395 | ||
| XM_005245515.4 | XP_005245572.1 | 3 | 1652 | |||
| GenBank mRNA | AK074830.1 | - | 7 | 2349 | ||
| AL136891.1 | - | 7 | 2359 | |||
| BC017306.2 | - | 7 | 2331 | 5088037 | ||
| BX537564.1 | - | 1 | 5432 | |||
| CR749209.1 | - | 3 | 5824 | |||

NUAK2
Genomic Map