siRNA Details
| Gene Symbol: NEK8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_178170.2 | NP_835464.1 | 15 | 2765 | ||
| XM_011524638.2 | XP_011522940.1 | 16 | 3008 | |||
| XM_011524640.2 | XP_011522942.1 | 16 | 2969 | |||
| XM_017024499.1 | XP_016879988.1 | 15 | 2814 | |||
| XM_017024500.1 | XP_016879989.1 | 15 | 2850 | |||
| GenBank mRNA | AY242354.1 | - | 15 | 2765 | ||

NEK8
Genomic Map