siRNA Details
Gene Symbol: NLRC3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_178844.3 | NP_849172.2 | 12, 13 | 2951 | ||
NR_075083.2 | 13, 14 | 3148 | ||||
XM_017023027.1 | XP_016878516.1 | 12, 13 | 2981 | |||
XM_017023028.1 | XP_016878517.1 | 13, 14 | 3155 | |||
XM_017023029.1 | XP_016878518.1 | 14, 15 | 3236 | |||
XM_017023030.1 | XP_016878519.1 | 14, 15 | 3297 | |||
XM_017023031.1 | XP_016878520.1 | 12, 13 | 3042 | |||
XM_017023032.1 | XP_016878521.1 | 14, 15 | 3998 | |||
XM_017023033.1 | XP_016878522.1 | 14, 15 | 3175 | |||
XM_017023034.1 | XP_016878523.1 | 13, 14 | 3094 | |||
XM_017023035.1 | XP_016878524.1 | 13, 14 | 3123 | |||
XM_017023036.1 | XP_016878525.1 | 12, 13 | 3682 | |||
XM_017023037.1 | XP_016878526.1 | 13, 14 | 2876 | |||
XM_017023038.1 | XP_016878527.1 | 13, 14 | 2849 | |||
XM_017023039.1 | XP_016878528.1 | 12, 13 | 2981 | |||
GenBank mRNA | AF501291.1 | - | 5, 6 | 428 | ||
AF501292.1 | - | 5, 6 | 428 | |||
AK074109.1 | - | 8, 9 | 829 | |||
AK090431.1 | - | 13, 14 | 3144 | |||
AY601811.1 | - | 12, 13 | 2938 | |||
FJ889357.1 | - | 9, 10 | 2527 |
