siRNA Details
| Gene Symbol: CDH9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_016279.3 | NP_057363.3 | 12 | 2550 | ||
| GenBank mRNA | AB035302.1 | - | 12 | 2535 | ||
| AK294183.1 | - | 5 | 1397 | |||
| AK307779.1 | - | 12 | 2551 | |||
| BC107594.1 | - | 12 | 2512 | 40011942 | ||
| BC113745.1 | - | 12 | 2512 | 40011941 | ||

CDH9
Genomic Map