siRNA Details
| Gene Symbol: SULT1C4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001321770.1 | NP_001308699.1 | 1, 2 | 604 | ||
| NM_006588.3 | NP_006579.2 | 1, 2 | 604 | |||
| GenBank mRNA | AK297851.1 | - | 1, 2 | 382 | ||
| AK298755.1 | - | 1, 2 | 545 | |||
| BC058861.1 | - | 1, 2 | 604 | |||
| BC125043.1 | - | 1, 2 | 330 | 40115974 | ||
| HY118496.1 | - | 21 | ||||

SULT1C4
Genomic Map