siRNA Details
| Gene Symbol: DTL | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001286229.1 | NP_001273158.1 | 2 | 440 | ||
| NM_001286230.1 | NP_001273159.1 | 2 | 440 | |||
| NM_016448.3 | NP_057532.3 | 3 | 566 | |||
| XM_011509614.1 | XP_011507916.1 | 3 | 270 | |||
| GenBank mRNA | AF195765.1 | - | 3 | 330 | ||
| AF345896.1 | - | 3 | 351 | |||
| AK000742.1 | - | 3 | 384 | |||
| AK027651.1 | - | 2 | 147 | |||
| AK292343.1 | - | 3 | 438 | |||
| BC033297.1 | - | 3 | 307 | 4342989 | ||
| BC033540.1 | - | 3 | 348 | 4826434 | ||
| DQ641253.1 | - | 3 | 228 | |||

DTL
Genomic Map