siRNA Details
Gene Symbol: PBDC1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_016500.3 | NP_057584.2 | 5 | 516 | ||
GenBank mRNA | BC001220.1 | - | 5 | 516 | 3528586 | |
BC051894.1 | - | 5 | 378 | 6528202 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: PBDC1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_016500.3 | NP_057584.2 | 5 | 516 | ||
GenBank mRNA | BC001220.1 | - | 5 | 516 | 3528586 | |
BC051894.1 | - | 5 | 378 | 6528202 |