siRNA Details
| Gene Symbol: DDX47 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_016355.3 | NP_057439.2 | 5 | 514 | ||
| NM_201224.1 | NP_957518.1 | 5 | 514 | |||
| GenBank mRNA | AF078843.1 | - | 5 | 471 | ||
| AF190165.1 | - | 5 | 478 | |||
| AJ276704.1 | - | 5 | 492 | |||
| AK054574.1 | - | 5 | 499 | |||
| AK222940.1 | - | 5 | 473 | |||
| AK225144.1 | - | 5 | 484 | |||
| AK302536.1 | - | 3 | 1010 | |||
| AK310501.1 | - | 5 | 544 | |||
| AL136666.1 | - | 5 | 518 | |||
| AL833469.1 | - | 4 | 314 | |||
| BC009379.2 | - | 5 | 477 | |||
| BC068009.1 | - | 5 | 496 | 6142837 | ||
| EF036507.1 | - | 5 | 461 | |||

DDX47
Genomic Map