siRNA Details
| Gene Symbol: TPRA1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001136053.2 | NP_001129525.1 | 9 | 1052 | ||
| NR_073377.1 | 8 | 991 | ||||
| XM_006713495.3 | XP_006713558.1 | 10 | 1233 | |||
| XM_006713496.2 | XP_006713559.1 | 9 | 910 | |||
| XM_006713497.3 | XP_006713560.1 | 10 | 1277 | |||
| XM_011512422.1 | XP_011510724.1 | 9 | 826 | |||
| XM_017005707.1 | XP_016861196.1 | 10 | 1016 | |||
| XM_017005708.1 | XP_016861197.1 | 10 | 1046 | |||
| XM_017005709.1 | XP_016861198.1 | 9 | 1086 | |||
| XM_017005710.1 | XP_016861199.1 | 9 | 925 | |||
| GenBank mRNA | AB037108.1 | - | 8 | 976 | ||
| BC017058.1 | - | 8 | 760 | 3904719 | ||
| BC050711.2 | - | 9 | 946 | 6168632 | ||
| CD580484.2 | - | 9 | 787 | |||

TPRA1
Genomic Map