siRNA Details
| Gene Symbol: TUBG2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001320509.1 | NP_001307438.1 | 12 | 1559 | ||
| NM_016437.2 | NP_057521.1 | 11 | 1532 | |||
| XM_011524614.2 | XP_011522916.1 | 10 | 1467 | |||
| XM_017024453.1 | XP_016879942.1 | 10 | 1447 | |||
| XM_017024454.1 | XP_016879943.1 | 11 | 2718 | |||
| XM_017024455.1 | XP_016879944.1 | 11 | 1546 | |||
| XM_017024456.1 | XP_016879945.1 | 11 | 2795 | |||
| XM_017024457.1 | XP_016879946.1 | 12 | 1360 | |||
| GenBank mRNA | AF225971.1 | - | 11 | 1525 | ||
| AK022324.1 | - | 11 | 1475 | |||
| AK225134.1 | - | 11 | 1491 | |||
| BC009670.2 | - | 11 | 1384 | 3891214 | ||
| BC051890.1 | - | 11 | 1475 | 5726689 | ||
| BC108739.1 | - | 11 | 1493 | 5242733 | ||
| BE727957.1 | - | 3 | 397 | |||

TUBG2
Genomic Map