siRNA Details
| Gene Symbol: TMEM8B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_016446.3 | NP_057530.2 | 11 | 2005 | ||
| XM_011517904.2 | XP_011516206.1 | 10 | 2388 | |||
| XM_011517905.1 | XP_011516207.1 | 11 | 2209 | |||
| GenBank mRNA | AF070572.1 | - | 9 | 1654 | ||
| AF188239.1 | - | 11 | 1966 | |||
| AK074844.1 | - | 9 | 2328 | |||
| BC098265.1 | - | 8 | 1548 | 40025067 | ||
| BC098296.1 | - | 8 | 1548 | 40025068 | ||
| BC099733.1 | - | 8 | 1548 | 40025069 | ||

TMEM8B
Genomic Map