siRNA Details
| Gene Symbol: TMEM14C | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001165258.1 | NP_001158730.1 | 6 | 874 | ||
| NM_016462.3 | NP_057546.1 | 6 | 860 | |||
| GenBank mRNA | AF151028.1 | - | 6 | 670 | ||
| AF164962.1 | - | 5 | 605 | |||
| BC002496.2 | - | 6 | 595 | 3050609 | ||
| BC010086.2 | - | 6 | 624 | 2907072 | ||

TMEM14C
Genomic Map