siRNA Details
| Gene Symbol: TMEM69 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_016486.3 | NP_057570.2 | 3 | 848 | ||
| GenBank mRNA | AF151063.1 | - | 3 | 827 | ||
| AK024682.1 | - | 3 | 842 | |||
| BC013608.1 | - | 3 | 830 | 3864518 | ||
| BC034147.1 | - | 3 | 828 | 4655713 | ||
| BC039718.2 | - | 2 | 751 | 5784175 | ||
| BC046130.1 | - | 3 | 811 | 5787422 | ||
| BC054041.1 | - | 1 | 562 | |||
| BC064539.1 | - | 3 | 831 | 5725426 | ||
| BC073836.1 | - | 2 | 748 | 5548608 | ||
| BC121117.2 | - | 3 | 790 | 40120483 | ||
| BC133692.1 | - | 3 | 789 | 40120480 | ||

TMEM69
Genomic Map