siRNA Details
| Gene Symbol: CLEC1B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001099431.1 | NP_001092901.1 | 3 | 462 | ||
| NM_016509.3 | NP_057593.3 | 4 | 561 | |||
| XM_005253382.4 | XP_005253439.1 | 6 | 933 | |||
| XM_011520685.2 | XP_011518987.1 | 5 | 842 | |||
| XM_011520686.2 | XP_011518988.1 | 4 | 470 | |||
| XM_017019395.1 | XP_016874884.1 | 5 | 917 | |||
| XM_017019396.1 | XP_016874885.1 | 3 | 269 | |||
| GenBank mRNA | AF124841.1 | - | 4 | 418 | ||
| AY358599.1 | - | 4 | 542 | |||
| BC029554.1 | - | 5 | 533 | 5269252 | ||
| BX647321.1 | - | 3 | 253 | |||

CLEC1B
Genomic Map