siRNA Details
| Gene Symbol: DCDC2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001195610.1 | NP_001182539.1 | 10 | 1471 | ||
| NM_016356.4 | NP_057440.2 | 9 | 1564 | |||
| GenBank mRNA | AB032980.2 | - | 9 | 1262 | ||
| AF181721.1 | - | 9 | 1564 | |||
| AK027036.1 | - | 9 | 1380 | |||
| BC050704.1 | - | 10 | 1471 | 5758845 | ||

DCDC2
Genomic Map