siRNA Details
| Gene Symbol: LUC7L3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_006107.3 | NP_006098.2 | 8 | 923 | ||
| NM_016424.4 | NP_057508.2 | 8 | 923 | |||
| XM_005257448.1 | XP_005257505.1 | 8 | 891 | |||
| XM_005257449.2 | XP_005257506.1 | 8 | 891 | |||
| XM_005257452.1 | XP_005257509.1 | 8 | 891 | |||
| XM_006721943.2 | XP_006722006.1 | 8 | 670 | |||
| GenBank mRNA | AF147357.1 | - | 3 | 289 | ||
| AK001925.1 | - | 8 | 857 | |||
| AK023672.1 | - | 8 | 885 | |||
| AK225980.1 | - | 8 | 891 | |||
| AK290540.1 | - | 8 | 890 | |||
| AK295983.1 | - | 7 | 792 | |||
| DQ013876.1 | - | 8 | 859 | |||

LUC7L3
Genomic Map