siRNA Details
| Gene Symbol: EMC4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001286420.1 | NP_001273349.1 | 4 | 672 | ||
| NM_016454.3 | NP_057538.1 | 5 | 833 | |||
| GenBank mRNA | AF151018.1 | - | 5 | 786 | ||
| AK075227.1 | - | 5 | 812 | |||
| AK290524.1 | - | 4 | 656 | |||
| BC002583.2 | - | 4 | 637 | 3161718 | ||
| DB506932.1 | - | 2 | 242 | |||

EMC4
Genomic Map