siRNA Details
| Gene Symbol: TMEM138 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_016464.4 | NP_057548.1 | 5 | 1166 | ||
| NR_028473.1 | 6 | 1049 | ||||
| XM_006718585.3 | XP_006718648.1 | 4 | 1404 | |||
| XM_006718586.2 | XP_006718649.1 | 5 | 803 | |||
| XM_006718588.3 | XP_006718651.1 | 5 | 661 | |||
| GenBank mRNA | AF151030.1 | - | 5 | 783 | ||
| AF151032.1 | - | 5 | 780 | |||
| AK027724.1 | - | 5 | 1315 | |||
| AL832879.1 | - | 3 | 3319 | |||
| BC005201.1 | - | 5 | 781 | 3684390 | ||

TMEM138
Genomic Map