siRNA Details
| Gene Symbol: OSER1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_016470.7 | NP_057554.4 | 4 | 1177 | ||
| XM_011528851.1 | XP_011527153.1 | 5 | 1652 | |||
| XM_011528853.2 | XP_011527155.1 | 4 | 1221 | |||
| XM_011528854.2 | XP_011527156.1 | 3 | 960 | |||
| XM_017027873.1 | XP_016883362.1 | 5 | 1717 | |||
| GenBank mRNA | AF151041.1 | - | 4 | 1061 | ||
| AF161517.1 | - | 4 | 1060 | |||
| AF217514.1 | - | 4 | 1045 | |||
| AK000476.1 | - | 4 | 1082 | |||
| AL133000.1 | - | 4 | 1062 | |||
| BC064973.1 | - | 4 | 1027 | 6151501 | ||

OSER1
Genomic Map