siRNA Details
| Gene Symbol: SHISA5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001272065.1 | NP_001258994.1 | 6 | 2144 | ||
| NM_001272066.1 | NP_001258995.1 | 6 | 2131 | |||
| NM_001272067.1 | NP_001258996.1 | 6 | 2326 | |||
| NM_001272068.1 | NP_001258997.1 | 7 | 2363 | |||
| NM_001272082.1 | NP_001259011.1 | 4 | 2020 | |||
| NM_001272083.1 | NP_001259012.1 | 3 | 2194 | |||
| NM_016479.4 | NP_057563.3 | 6 | 2165 | |||
| GenBank mRNA | AF520698.1 | - | 6 | 2144 | ||

SHISA5
Genomic Map