siRNA Details
| Gene Symbol: C6orf203 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001142468.2 | NP_001135940.1 | 3 | 1152 | ||
| NM_001142470.2 | NP_001135942.1 | 4 | 1235 | |||
| NM_016487.4 | NP_057571.1 | 3 | 626 | |||
| XM_011535876.2 | XP_011534178.1 | 4 | 730 | |||
| XM_011535877.2 | XP_011534179.1 | 4 | 678 | |||
| XM_017010919.1 | XP_016866408.1 | 3 | 625 | |||
| XM_017010920.1 | XP_016866409.1 | 3 | 1118 | |||
| GenBank mRNA | AF151064.1 | - | 3 | 582 | ||
| AK091564.1 | - | 3 | 1111 | |||
| BC010899.2 | - | 2 | 528 | 4078296 | ||

C6orf203
Genomic Map