siRNA Details
| Gene Symbol: RNF181 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_016494.3 | NP_057578.1 | 4 | 442 | ||
| XM_005264359.3 | XP_005264416.1 | 3 | 929 | |||
| GenBank mRNA | AF151072.1 | - | 4 | 422 | ||
| AK222700.1 | - | 4 | 408 | |||
| BC002803.2 | - | 4 | 417 | 3636703 | ||
| CB131242.1 | - | 4 | 458 | |||
| CR457165.1 | - | 4 | 392 | |||

RNF181
Genomic Map