siRNA Details
| Gene Symbol: TMEM216 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001173990.2 | NP_001167461.1 | 5 | 876 | ||
| NM_001173991.2 | NP_001167462.1 | 5 | 885 | |||
| NM_016499.5 | NP_057583.2 | 5 | 866 | |||
| XM_005274039.3 | XP_005274096.1 | 6 | 1706 | |||
| XM_017017882.1 | XP_016873371.1 | 5 | 1563 | |||
| GenBank mRNA | AF151078.1 | - | 5 | 627 | ||
| AK303687.1 | - | 5 | 876 | |||
| BC011010.1 | - | 5 | 656 | 4286625 | ||
| KU159424.1 | - | 5 | 663 | |||

TMEM216
Genomic Map