siRNA Details
| Gene Symbol: ZCCHC17 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001282566.1 | NP_001269495.1 | 6 | 713 | ||
| NM_001282567.1 | NP_001269496.1 | 6 | 653 | |||
| NM_001282568.1 | NP_001269497.1 | 7 | 834 | |||
| NM_001282569.1 | NP_001269498.1 | 8 | 766 | |||
| NM_001282570.1 | NP_001269499.1 | 6 | 575 | |||
| NM_001282571.1 | NP_001269500.1 | 6 | 595 | |||
| NM_016505.3 | NP_057589.2 | 7 | 696 | |||
| XM_006710681.3 | XP_006710744.1 | 8 | 840 | |||
| XM_011541567.2 | XP_011539869.1 | 7 | 712 | |||
| GenBank mRNA | AF151077.1 | - | 7 | 625 | ||
| AF151085.1 | - | 7 | 652 | |||
| AF247661.2 | - | 6 | 565 | |||
| AK001929.1 | - | 7 | 646 | |||
| AK022506.1 | - | 7 | 683 | |||
| AK024049.1 | - | 7 | 672 | |||
| AK302248.1 | - | 8 | 716 | |||
| AK302343.1 | - | 6 | 599 | |||
| BC000685.2 | - | 7 | 734 | |||
| BC007446.2 | - | 7 | 632 | 3009823 | ||
| BC050609.1 | - | 7 | 636 | 6527025 | ||
| BP309837.1 | - | 6 | 536 | |||
| BP371153.1 | - | 6 | 531 | |||

ZCCHC17
Genomic Map