siRNA Details
| Gene Symbol: AMBN | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_016519.5 | NP_057603.1 | 7 | 647 | ||
| GenBank mRNA | AF219994.2 | - | 7 | 611 | ||
| AF263464.1 | - | 7 | 621 | |||
| AJ537437.1 | - | 7 | 621 | |||
| AK314918.1 | - | 7 | 582 | |||
| BC106931.1 | - | 7 | 582 | 40033703 | ||

AMBN
Genomic Map