siRNA Details
| Gene Symbol: KLRF1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001291822.1 | NP_001278751.1 | 5 | 693 | ||
| NM_001291823.1 | NP_001278752.1 | 3 | 440 | |||
| NM_016523.2 | NP_057607.1 | 6 | 843 | |||
| NR_120305.1 | 4 | 553 | ||||
| XM_017019414.1 | XP_016874903.1 | 4 | 709 | |||
| GenBank mRNA | AF175206.1 | - | 6 | 843 | ||
| AF175207.1 | - | 5 | 693 | |||
| AF267244.1 | - | 4 | 553 | |||
| AF267245.1 | - | 3 | 440 | |||
| AJ305370.1 | - | 6 | 826 | |||
| BC096734.1 | - | 4 | 550 | |||
| BC098166.1 | - | 5 | 682 | 40015535 | ||
| BC098257.1 | - | 5 | 663 | 40015534 | ||
| BC098354.1 | - | 6 | 804 | 40015531 | ||

KLRF1
Genomic Map