siRNA Details
| Gene Symbol: COA4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_016565.2 | NP_057649.2 | 2 | 299 | ||
| XM_017017883.1 | XP_016873372.1 | 2 | 615 | |||
| XM_017017884.1 | XP_016873373.1 | 3 | 452 | |||
| GenBank mRNA | AF242180.1 | - | 1 | 352 | ||
| AK000434.1 | - | 2 | 299 | |||
| AK314104.1 | - | 2 | 615 | |||
| AL359618.1 | - | 2 | 279 | |||
| BC106036.1 | - | 2 | 315 | 4445548 | ||

COA4
Genomic Map