siRNA Details
| Gene Symbol: ERGIC2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_016570.2 | NP_057654.2 | 2, 3 | 241 | ||
| GenBank mRNA | AF183410.1 | - | 2, 3 | 192 | ||
| AF216751.1 | - | 2, 3 | 196 | |||
| AF302767.1 | - | 2, 3 | 194 | |||
| AK074520.1 | - | 2, 3 | 216 | |||
| AK222799.1 | - | 2, 3 | 181 | |||
| AL834128.2 | - | 2, 3 | 182 | |||
| BC000887.1 | - | 2, 3 | 194 | 3462408 | ||
| BC107794.1 | - | 2, 3 | 198 | 30319807 | ||
| BF681475.1 | - | 267 | ||||
| BI547753.1 | - | 2, 3 | 245 | |||

ERGIC2
Genomic Map