siRNA Details
| Gene Symbol: PART1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_024617.1 | 1 | 345 | |||
| NR_028508.1 | 1 | 126 | ||||
| NR_028509.1 | 1 | 126 | ||||
| GenBank mRNA | AF163475.1 | - | 1 | 126 | ||
| AK312071.1 | - | 1 | 348 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

PART1| Gene Symbol: PART1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_024617.1 | 1 | 345 | |||
| NR_028508.1 | 1 | 126 | ||||
| NR_028509.1 | 1 | 126 | ||||
| GenBank mRNA | AF163475.1 | - | 1 | 126 | ||
| AK312071.1 | - | 1 | 348 | |||