siRNA Details
| Gene Symbol: FAM13B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AF251038.1 | - | 3 | 551 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

FAM13B| Gene Symbol: FAM13B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AF251038.1 | - | 3 | 551 | ||