siRNA Details
| Gene Symbol: DNAJC27 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001198559.1 | NP_001185488.1 | 3, 4 | 438 | ||
| NM_016544.2 | NP_057628.1 | 3, 4 | 438 | |||
| XM_011532902.1 | XP_011531204.1 | 3, 4 | 413 | |||
| XM_017004269.1 | XP_016859758.1 | 3, 4 | 469 | |||
| XM_017004270.1 | XP_016859759.1 | 3, 4 | 329 | |||
| GenBank mRNA | AF178983.1 | - | 3, 4 | 272 | ||
| AK294726.1 | - | 3, 4 | 222 | |||
| AY094594.1 | - | 3, 4 | 248 | |||
| BC034049.1 | - | 3, 4 | 299 | 4815297 | ||

DNAJC27
Genomic Map