siRNA Details
| Gene Symbol: RASL12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001307930.1 | NP_001294859.1 | 4 | 692 | ||
| NM_016563.3 | NP_057647.1 | 5 | 749 | |||
| XM_011521660.2 | XP_011519962.1 | 6 | 562 | |||
| GenBank mRNA | AF233588.1 | - | 5 | 588 | ||
| AK296261.1 | - | 4 | 502 | |||
| AK300536.1 | - | 5 | 532 | |||
| AK314914.1 | - | 5 | 557 | |||
| BC053734.1 | - | 5 | 597 | 5588236 | ||

RASL12
Genomic Map