siRNA Details
| Gene Symbol: GMPR2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001002000.2 | NP_001002000.1 | 3, 4 | 664 | ||
| NM_001002001.2 | NP_001002001.1 | 3, 4 | 605 | |||
| NM_001002002.2 | NP_001002002.1 | 3, 4 | 344 | |||
| NM_001283022.1 | NP_001269951.1 | 2, 3 | 585 | |||
| NM_016576.4 | NP_057660.2 | 2, 3 | 585 | |||
| XM_005267740.3 | XP_005267797.1 | 3, 4 | 470 | |||
| XM_005267741.3 | XP_005267798.1 | 3, 4 | 529 | |||
| XM_005267742.3 | XP_005267799.1 | 3, 4 | 348 | |||
| GenBank mRNA | AB032903.1 | - | 2, 3 | 211 | ||
| AF086919.1 | - | 3, 4 | 374 | |||
| AF135159.1 | - | 3, 4 | 550 | |||
| AF419346.1 | - | 3, 4 | 454 | |||
| BC003053.1 | - | 2, 3 | 534 | 3546620 | ||
| BC008021.1 | - | 2, 3 | 533 | 3537989 | ||
| BC009832.2 | - | 3, 4 | 262 | 3958714 | ||
| BC093039.1 | - | 3, 4 | 403 | 30347740 | ||
| BI259066.1 | - | 2, 3 | 547 | |||
| BP347823.1 | - | 3, 4 | 344 | |||
| BX161436.1 | - | 3, 4 | 585 | |||
| CR457156.1 | - | 2, 3 | 203 | |||

GMPR2
Genomic Map