siRNA Details
| Gene Symbol: THEG | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_016585.4 | NP_057669.1 | 1 | 330 | ||
| NM_199202.2 | NP_954672.1 | 1 | 330 | |||
| XM_011528049.2 | XP_011526351.1 | 1 | 1009 | |||
| XM_011528050.2 | XP_011526352.1 | 1 | 1008 | |||
| XM_011528051.2 | XP_011526353.1 | 1 | 1010 | |||
| XM_011528052.2 | XP_011526354.1 | 1 | 1008 | |||
| GenBank mRNA | AB033129.1 | - | 1 | 330 | ||
| AF268610.1 | - | 1 | 296 | |||
| BC028574.2 | - | 1 | 309 | 4821678 | ||

THEG
Genomic Map