siRNA Details
| Gene Symbol: NRN1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001278710.1 | NP_001265639.1 | 4 | 474 | ||
| NM_001278711.1 | NP_001265640.1 | 4 | 713 | |||
| NM_016588.2 | NP_057672.1 | 3 | 972 | |||
| XM_006715106.3 | XP_006715169.1 | 5 | 1363 | |||
| XM_006715107.3 | XP_006715170.1 | 3 | 1153 | |||
| GenBank mRNA | AF114833.1 | - | 3 | 503 | ||
| AF136631.1 | - | 3 | 489 | |||
| AJ420483.1 | - | 3 | 446 | |||
| AK093824.1 | - | 3 | 1167 | |||
| AK314095.1 | - | 3 | 525 | |||
| AL541038.3 | - | 3 | 557 | |||
| BC002683.2 | - | 3 | 450 | 3605775 | ||
| BC042019.1 | - | 3 | 973 | 5312094 | ||
| BF970432.1 | - | 3 | 365 | |||
| BX340783.2 | - | 4 | 709 | |||
| DB013243.1 | - | 4 | 474 | |||
More Information
Other Species Targeted
This siRNA also targets the following transcripts in Mouse: NM_153529, BC035531, AK003046, AK161859.

NRN1
Genomic Map