siRNA Details
Gene Symbol: PART1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_024617.1 | 1 | 469 | |||
NR_028508.1 | 1 | 250 | ||||
NR_028509.1 | 1 | 250 | ||||
GenBank mRNA | AF163475.1 | - | 1 | 250 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: PART1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_024617.1 | 1 | 469 | |||
NR_028508.1 | 1 | 250 | ||||
NR_028509.1 | 1 | 250 | ||||
GenBank mRNA | AF163475.1 | - | 1 | 250 |