siRNA Details
| Gene Symbol: PART1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_028508.1 | 2 | 1203 | |||
| GenBank mRNA | AF163475.1 | - | 2 | 1203 | ||
| AL050198.1 | - | 2 | 947 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

PART1| Gene Symbol: PART1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_028508.1 | 2 | 1203 | |||
| GenBank mRNA | AF163475.1 | - | 2 | 1203 | ||
| AL050198.1 | - | 2 | 947 | |||