siRNA Details
| Gene Symbol: UFM1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001286703.1 | NP_001273632.1 | 6 | 2048 | ||
| NM_001286704.1 | NP_001273633.1 | 6 | 2226 | |||
| NM_001286705.1 | NP_001273634.1 | 7 | 2090 | |||
| NM_001286706.1 | NP_001273635.1 | 6 | 2724 | |||
| NM_016617.3 | NP_057701.1 | 6 | 2054 | |||
| NR_104584.1 | 7 | 2115 | ||||
| NR_104585.1 | 5 | 2014 | ||||
| GenBank mRNA | AF208844.1 | - | 6 | 1944 | ||
| AK095490.1 | - | 6 | 1974 | |||
| CB070175.1 | - | 1 | 500 | |||

UFM1
Genomic Map