siRNA Details
| Gene Symbol: CYP4F2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001082.4 | NP_001073.3 | 13 | 1904 | ||
| GenBank mRNA | AK290790.1 | - | 13 | 1858 | ||
| D26480.1 | - | 13 | 1849 | |||
| U02388.2 | - | 13 | 1867 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

CYP4F2| Gene Symbol: CYP4F2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001082.4 | NP_001073.3 | 13 | 1904 | ||
| GenBank mRNA | AK290790.1 | - | 13 | 1858 | ||
| D26480.1 | - | 13 | 1849 | |||
| U02388.2 | - | 13 | 1867 | |||