siRNA Details
| Gene Symbol: KLHL3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001257194.1 | NP_001244123.1 | 3, 4 | 868 | ||
| NM_017415.2 | NP_059111.2 | 3, 4 | 693 | |||
| GenBank mRNA | AB032955.1 | - | 3, 4 | 463 | ||
| AF208068.1 | - | 3, 4 | 377 | |||
| AF208070.1 | - | 3, 4 | 868 | |||
| AK314707.1 | - | 3, 4 | 434 | |||
| BC034035.1 | - | 3, 4 | 692 | 4817650 | ||

KLHL3
Genomic Map