siRNA Details
| Gene Symbol: SPA17 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_017425.3 | NP_059121.1 | 3 | 333 | ||
| XM_011542870.2 | XP_011541172.1 | 3 | 523 | |||
| GenBank mRNA | AF334735.1 | - | 3 | 332 | ||
| AK311806.1 | - | 3 | 276 | |||
| BC019224.1 | - | 3 | 265 | |||
| BC032457.2 | - | 2 | 225 | 5171849 | ||
| DA511916.1 | - | 3 | 382 | |||
| KU257683.1 | - | 2 | 197 | |||

SPA17
Genomic Map