siRNA Details
| Gene Symbol: OTUD4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001102653.1 | NP_001096123.1 | 3, 4 | 245 | ||
| NM_017493.6 | NP_059963.1 | 2, 3 | 143 | |||
| XM_005263079.4 | XP_005263136.1 | 3, 4 | 348 | |||
| XM_011532040.2 | XP_011530342.2 | 3, 4 | 349 | |||
| XM_011532041.2 | XP_011530343.2 | 3, 4 | 349 | |||
| XM_011532042.2 | XP_011530344.1 | 3, 4 | 270 | |||
| GenBank mRNA | AK302581.1 | - | 3, 4 | 207 | ||
| BC118572.1 | - | 2, 3 | 139 | 40036249 | ||
| BC118653.1 | - | 2, 3 | 139 | 40036248 | ||
| DB446926.2 | - | 3, 4 | 247 | |||
| X68242.1 | - | 3, 4 | 234 | |||
More Information
Other Species Targeted
This siRNA also targets the following transcripts in Mouse: NM_001081164, NM_001256033, BC138373, BC145259, AW494258.

OTUD4
Genomic Map