siRNA Details
Gene Symbol: DNAH3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017539.2 | NP_060009.1 | 1 | 104 | ||
XM_017023432.1 | XP_016878921.1 | 1 | 200 | |||
GenBank mRNA | AF494040.1 | - | 1 | 104 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: DNAH3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017539.2 | NP_060009.1 | 1 | 104 | ||
XM_017023432.1 | XP_016878921.1 | 1 | 200 | |||
GenBank mRNA | AF494040.1 | - | 1 | 104 |