siRNA Details
| Gene Symbol: EPDR1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001242946.1 | NP_001229875.2 | 1 | 252 | ||
| NM_017549.4 | NP_060019.2 | 1 | 252 | |||
| GenBank mRNA | AY027862.2 | - | 1 | 252 | ||
| BC018299.1 | - | 1 | 144 | 3461888 | ||

EPDR1
Genomic Map